Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TGF beta induced factor 2 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$343.50 - $573.00
Specifications
Antigen | TGF beta induced factor 2 |
---|---|
Dilution | Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml |
Applications | Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
TGF beta induced factor 2 Polyclonal antibody specifically detects TGF beta induced factor 2 in Human samples. It is validated for ImmunofluorescenceSpecifications
TGF beta induced factor 2 | |
Immunofluorescence | |
Unconjugated | |
Rabbit | |
Angiogenesis, Cancer, Core ESC Like Genes, Stem Cell Markers | |
PBS, pH 7.2, 40% glycerol | |
60436 | |
IgG | |
Affinity purified |
Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml | |
Polyclonal | |
Purified | |
RUO | |
Human | |
homeobox protein TGIF2, TGF(beta)-induced transcription factor 2, TGF-beta-induced transcription factor 2,5'-TG-3'-interacting factor 2,5'-TG-3' interacting factor 2, TGFB-induced factor 2, TGFB-induced factor 2 (TALE family homeobox), TGFB-induced factor homeobox 2, transcription growth factor-beta-induced factor 2 | |
This antibody was developed against Recombinant Protein corresponding to amino acids: RAAEMELQKQQDPSLPLLHTPIPLVSE | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title