Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TGF-beta Mouse anti-Human, Mouse, Rat, Bovine, Canine, Hamster, Monkey, Clone: 1D11.16.8, Novus Biologicals™

Mouse Monoclonal Antibody

$138.37 - $352.89

Specifications

Antigen TGF-beta
Clone 1D11.16.8
Concentration 0.2 mg/ml
Dilution Western Blot 1-2 ug/ml, ELISA 0.1-0.4 ug/ml, Immunocytochemistry/Immunofluorescence 8-25 ug/ml, Functional Assay
Applications Western Blot, ELISA, Immunofluorescence, Functional Assay
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122909
View Documents
Novus Biologicals
NBP30771810UG
100 μg
Each for $352.89
Add to cart
 
NB122910
View Documents
Novus Biologicals
NBP30771820UG
20 μg
Each for $138.37
Add to cart
 
Description

Description

TGF-beta Monoclonal antibody specifically detects TGF-beta in Human, Mouse, Rat, Bovine, Canine, Hamster, Monkey samples. It is validated for Western Blot, ELISA, Immunofluorescence, Functional Assay
Specifications

Specifications

TGF-beta
0.2 mg/ml
Western Blot, ELISA, Immunofluorescence, Functional Assay
Purified
RUO
10 mM PBS with 0.05% BSA
7040
IgG1 Kappa
Primary
Store at 4°C.
1D11.16.8
Western Blot 1-2 ug/ml, ELISA 0.1-0.4 ug/ml, Immunocytochemistry/Immunofluorescence 8-25 ug/ml, Functional Assay
Unconjugated
Mouse
Human, Mouse, Rat, Bovine, Canine, Hamster, Monkey
CED, DPD1, LAP, TGFB, TGFB1, TGFbeta, transforming growth factor beta 1
Bovine TGF-beta protein
Monoclonal
Protein A or G purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit