Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TGF-beta Mouse anti-Human, Mouse, Rat, Bovine, Canine, Hamster, Monkey, Clone: TGFB/510, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP30770810UG

 View more versions of this product

Catalog No. NB122889


Add to cart

Description

Description

TGF-beta Monoclonal specifically detects TGF-beta in Human, Mouse, Rat, Bovine, Canine, Hamster, Monkey samples. It is validated for ELISA, Functional Assay.
Specifications

Specifications

TGF-beta
TGFB/510
Unconjugated
10 mM PBS with 0.05% BSA
CED, DPD1, LAP, TGFB, TGFB1, TGFbeta, transforming growth factor beta 1
Full length recombinant human TGF-beta protein
Protein A or G purified
RUO
Primary
7040
ELISA, Functional Assay
0.2 mg/ml
ELISA 0.1-0.4 ug/ml, Functional Assay
Purified
Mouse
IgG1 Kappa
100 μg
Store at 4°C.
Monoclonal
Human, Mouse, Rat, Bovine, Canine, Hamster, Monkey
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit