Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TGF-beta RII, Goat anti-Mouse, Alexa Fluor 700, Polyclonal, R&D Systems™

Mouse Monoclonal Antibody

$194.12 - $370.59

Specifications

Host Species Goat
Conjugate Alexa Fluor 700
Isotype IgG
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse TGF-beta RII and S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse TGF-beta RII Ile24-Asp184 Accession # Q62312
Antigen TGF-beta RII
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
FAB532N025
View Documents
R&D Systems
FAB532N025
25 Tests
Each for $194.12
Add to cart
 
FAB532N100
View Documents
R&D Systems
FAB532N100
100 Tests
Each for $370.59
Add to cart
 
Description

Description

TGF-beta RII Polyclonal specifically detects TGF-beta RII in Mouse samples. It is validated for Flow Cytometry.
Specifications

Specifications

Goat
IgG
TGF-beta RII
Affinity Purified
RUO
Detects mouse TGF-beta RII in direct ELISAs and Western blots. In direct ELISAs, approximately 5% cross-reactivity with recombinant human (rh) TGF-beta RII is observed and less than 1% cross-reactivity with recombinant mouse TGF-beta RI and rhTGF-beta RIII is observed.
7048
Antigen Affinity-purified
Alexa Fluor 700
Mouse myeloma cell line NS0-derived recombinant mouse TGF-beta RII and S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse TGF-beta RII Ile24-Asp184 Accession # Q62312
Polyclonal
Primary
Flow Cytometry
Q62312
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit