Learn More
TGF-beta RIII Mouse anti-Human, Mouse, DyLight 488, Clone: 1C5H11, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237418G
Catalog No. NBP237418G
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
TGF-beta RIII
1C5H11
Western Blot, ELISA
Purified
TGFBR3
Purified recombinant fragment of human TGFBR3 (AA: 147-328 ) expressed in E. Coli.
Protein A or G purified
Angiogenesis
Primary
7049
ELISA, Western Blot
DyLight 488
50mM Sodium Borate with 0.05% Sodium Azide
betaglycan, betaglycan proteoglycan, BGCAN, TGF-beta receptor type 3, TGF-beta receptor type III, TGFR-3, Transforming growth factor beta receptor III, transforming growth factor beta receptor type 3, transforming growth factor, beta receptor III, transforming growth factor, beta receptor III (betaglycan, 300kDa)
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Human, Mouse