Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TGF-beta RIII Mouse anti-Human, Mouse, FITC, Clone: 1C5H11, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237418F

Catalog No. NBP237418F

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

TGF-beta RIII
1C5H11
ELISA
Purified
TGFBR3
Purified recombinant fragment of human TGFBR3 (AA: 147-328 ) expressed in E. Coli.
Protein A or G purified
Angiogenesis
Primary
7049
ELISA
FITC
PBS with 0.05% Sodium Azide
betaglycan, betaglycan proteoglycan, BGCAN, TGF-beta receptor type 3, TGF-beta receptor type III, TGFR-3, Transforming growth factor beta receptor III, transforming growth factor beta receptor type 3, transforming growth factor, beta receptor III, transforming growth factor, beta receptor III (betaglycan, 300kDa)
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit