Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TGFA Mouse anti-Human, Clone: 1E8-G6, Abnova™

Mouse monoclonal antibody raised against synthetic peptide of human TGFA.

Manufacturer:  Abnova Corporation MAB13444

Catalog No. 89-935-000


Add to cart

Description

Description

Transforming growth factors (TGFs) are biologically active polypeptides that reversibly confer the transformed phenotype on cultured cells. TGF-alpha shows about 40% sequence homology with epidermal growth factor (EGF; MIM 131530) and competes with EGF for binding to the EGF receptor (MIM 131550), stimulating its phosphorylation and producing a mitogenic response.[supplied by OMIM]

Sequence: PPVAAAVVSH
Specifications

Specifications

TGFA
1E8-G6
Mouse monoclonal antibody raised against synthetic peptide of human TGFA.
In 10mM PBS (0.05% sodium azide).
TFGA
Mouse
IgM, kappa
100 ug
Store at 4°C.
Monoclonal
This monoclonal antibody reacts with the TGFA and shows no cross-reaction with EGF and the neuropeptide synenkephalin.
Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed)
Unconjugated
Flow Cytometry (0.5-1 ug/106 cells in 0.1 mL) Immunofluorescence (1-2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (2-4 ug/mL) The optimal working dilution should be determined by the end user.
Liquid
TGFA
A synthetic peptide corresponding to amino acids 34-43 of human TGFA.
PEG precipitation
RUO
Primary
7039
Human
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit