Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TGFA Mouse anti-Human, Clone: TGFA/1119, Abnova™

Mouse Monoclonal Antibody

Manufacturer:  Abnova Corporation MAB14615

Catalog No. 89-034-698


Add to cart

Description

Description

Mouse monoclonal antibody raised against full length recombinant human TGFA.

  • High specificity
  • High reactivity
Specifications

Specifications

TGFA
TGFA/1119
Mouse monoclonal antibody raised against full length recombinant human TGFA.
In 10 mM PBS.
Liquid
TGFA
Recombinant protein corresponding to full length human TGFA.
Protein A/G purification
RUO
Primary
7039
Human
Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed)
Unconjugated
Flow Cytometry (0.5-1 ug/million cells in 0.1 mL) Immunofluorescence (1-2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (2-4 ug/mL) The optimal working dilution should be determined by the end user.
P01135
TFGA
Mouse
IgG1, kappa
100 ug
Store at -20 to -80°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
This antibody reacts with the C-terminus of TGFA and shows no cross-reaction with EGF and the neuropeptide synenkephalin.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only (RUO)