Learn More
Abnova™ TGFB1 (Bovine) ELISA Kit
Sandwich enzyme-linked immunosorbent assay
Manufacturer:  Abnova™ KA5280
Catalog No. 89-335-314
Add to cart
Description
Description
TGFB1 (Bovine) ELISA Kit is a sandwich enzyme-linked immunosorbent assay for the quantitative measurement of bovine TGFB1.
Specifications
Specifications
Bovine
Quantitative
RUO
1 Kit
transforming growth factor, beta 1
282089
Store at 4°C for 6 months, at -20°C for 12 months. Avoid multiple freeze-thaw cycles.
Bovine