Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TGFBR3L Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP257581

Catalog No. NBP257581


Add to cart

Description

Description

TGFBR3L Polyclonal antibody specifically detects TGFBR3L in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

TGFBR3L
Unconjugated
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide
TGF-Beta Receptor Type III-Like Protein, TGF-Beta Receptor Type-3-Like Protein, TGFR-3L, Transforming Growth Factor Beta Receptor 3 Like, Transforming Growth Factor, Beta Receptor III-Like, Transforming Growth Factor-Beta Receptor Type 3-Like Protein, Transforming Growth Factor-Beta Receptor Type III-Like Protein
Rabbit
IgG
100 ul
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin)
Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 1-4 ug/ml, Immunohistochemistry-Paraffin 1:50 - 1:200
Affinity Purified
TGFBR3L
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:FPGGLKGSARFLSFGPPFPAPPAPPFPAAPGPWLRRPLFSLKLSDTEDVFP
Affinity purified
RUO
Primary
100507588
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only