Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TGFBRAP1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548571

Catalog No. PA548571

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Plays a role in the TGF-beta/activin signaling pathway. It associates with inactive heteromeric TGF-beta and activin receptor complexes, mainly through the type II receptor, and is released upon activation of signaling. May recruit SMAD4 to the vicinity of the receptor complex and facilitate its interaction with receptor-regulated Smads, such as SMAD2.
Specifications

Specifications

TGFBRAP1
Polyclonal
PBS with 0.09% sodium azide
Q8WUH2
Transforming growth factor-beta receptor-associated protein 1, TGF-beta receptor-associated protein 1, TRAP-1, TRAP1, TGFBRAP1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
9392
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
TGFBRAP1
Liquid
TGFBRAP1
KLH conjugated synthetic peptide between 499-528 amino acids from the C-terminal region of human TGFBRAP1.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit