Learn More
TGFBRAP1 Rabbit anti-Human, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 201531AP
Catalog No. 50-173-2736
Add to cart
Description
Description
Plays a role in the TGF-beta/activin signaling pathway. It associates with inactive heteromeric TGF-beta and activin receptor complexes, mainly through the type II receptor, and is released upon activation of signaling. May recruit SMAD4 to the vicinity of the receptor complex and facilitate its interaction with receptor-regulated Smads, such as SMAD2.
Specifications
Specifications
TGFBRAP1
0.15 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q8WUH2
TGFBRAP1, TRAP 1, TRAP1
Rabbit
IgG
150 μL
-20°C
Primary
9392
Immunohistochemistry (Paraffin), Immunoprecipitation
Unconjugated
TGFBRAP1
Liquid
TGFBRAP1
TGFBRAP1 Fusion Protein Ag13698
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human