Learn More
TGM1 Mouse anti-Human, Clone: 6.4_2G6, Abnova™
Mouse monoclonal antibody raised against human TGM1.
Manufacturer:  Abnova Corporation MAB12758
Catalog No. 89-935-002
Add to cart
Description
Description
The protein encoded by this gene is a membrane protein that catalyzes the addition of an alkyl group from an akylamine to a glutamine residue of a protein, forming an alkylglutamine in the protein. This protein alkylation leads to crosslinking of proteins and catenation of polyamines to proteins. This gene contains either one or two copies of a 22 nt repeat unit in its 3' UTR. Mutations in this gene have been associated with autosomal recessive lamellar ichthyosis (LI) and nonbullous congenital ichthyosiform erythroderma (NCIE). [provided by RefSeq]
Specifications
Specifications
TGM1
6.4_2G6
Mouse monoclonal antibody raised against human TGM1.
In PBS (0.02% sodium azide).
ICR2/KTG/LI/LI1/TGASE/TGK
Mouse
IgG2a
50 ug
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
Immunocytochemistry, Immunohistochemistry (PFA fixed), Immunoprecipitation, Western Blot
Unconjugated
Immunocytochemistry (10 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) Immunoprecipitation Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user.
Liquid
TGM1
Human HeLa cells
Protein A/G purification
RUO
Primary
7051