Learn More
TGM4 Rabbit anti-Human, Mouse, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 158471AP
Catalog No. 50-173-0525
Add to cart
Description
Description
TGM4 is associated with the mammalian reproductive process. TGM4 catalyzes the cross-linking of proteins and the conjugation of polyamines to specific proteins in the seminal tract.
Specifications
Specifications
TGM4
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P49221, Q8BZH1
Fibrinoligase, hTGP, Prostate transglutaminase, TG(P), TGase 4, TGase P, TGM4, TGP, Transglutaminase 4, transglutaminase 4 (prostate), Transglutaminase P
Rabbit
IgG
150 μL
-20°C
Primary
331046, 7047
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
TGM4
Liquid
Tgm4
TGM4 Fusion Protein Ag8608
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse