Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TGM5 Polyclonal Antibody
TGM5 Polyclonal Antibody
Supplier: Thermo Scientific PA572718
Description
TGM5 Polyclonal Antibody for Western Blot, IHC (P), ELISA

Specifications
TGM5 | |
Polyclonal | |
Unconjugated | |
Tgm5 | |
2310007C07Rik; MGC141907; protein-glutamine gamma-glutamyltransferase 5; PSS2; TG(X); TGase 5; TGase X; TGASE5; TGase-5; TGASEX; Tgm5; TGM6; TGMX; TGX; transglutaminase 5; transglutaminase V; transglutaminase X; Transglutaminase-5 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
691929, 74176, 9333 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
O43548, Q9D7I9 | |
Tgm5 | |
TGM5 antibody for a 19 amino acid synthetic peptide near the carboxy terminus of human TGM5. The immunogen is within the last 50 amino acids of TGM5. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction