Learn More
TGN38 Rabbit anti-Human, Mouse, Rat, Monkey, Janelia Fluor 646, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus Biologicals NBP103495JF646
Catalog No. NBP13495646
Add to cart
Description
Description
TGN38 Polyclonal antibody specifically detects TGN38 in Human, Mouse, Rat, Monkey samples. It is validated for Immunohistochemistry, Immunocytochemistry, Immunofluorescence.
Specifications
Specifications
TGN38
Janelia Fluor 646
50mM Sodium Borate with 0.05% Sodium Azide
TGN46TGN38 homolog, TGN48MGC14722, TGN5148, 51kD isoforms), trans-golgi network protein 2, Trans-Golgi network protein TGN51
Rabbit
IgG
0.1mL
Store at 4°C in the dark.
Polyclonal
TGN38 (or TGN46, TGN48, TGN51 in human).
Immunocytochemistry, Immunofluorescence, Immunohistochemistry
Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Purified
TGOLN2
A synthetic peptide from the cytoplasmic part of mouse TGN38 conjugated to an immunogenic carrier protein was used as the immunogen.
IgG purified
Golgi Apparatus Markers, Trans golgi Network Markers
Primary
10618
Human, Monkey, Mouse, Rat