Alfa Aesar™ Thallium(I) 2-ethylhexanoate, 99.8% (metals basis)

Manufacturer: Alfa Aesar™ 3665909

Quantity 10g

 View more versions of this product

Catalog No. AA3665909

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 210578-56-6
    Formula Weight 347.59
    MDL Number MFCD00070466
    UN Number UN1707
    Chemical Name or Material Thallium(I) 2-ethylhexanoate

    Specifications

    Formula Weight 347.59
    Health Hazard 3 GHS P Statement
    P260-P301+P310-P304+P340-P320-P330-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00070466
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H300-H330-H373
    Fatal if swallowed.
    Fatal if inhaled.
    May cause damage to organs through prolonged or repeated exposure.
    Physical Form Solid
    CAS 210578-56-6
    UN Number UN1707
    Quantity 10g
    DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: THALLIUM COMPOUNDS, N.O.S.
    Chemical Name or Material Thallium(I) 2-ethylhexanoate
    Assay 99.8%, (metals basis)

    GHS H Statement
    H300-H330-H373
    Fatal if swallowed.
    Fatal if inhaled.
    May cause damage to organs through prolonged or repeated exposure.

    GHS P Statement
    P260-P301+P310-P304+P340-P320-P330-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit