Learn More
Thallium(I) acetate, 99.995% (metals basis), Thermo Scientific Chemicals
Description
Thallium(I) acetate is used as a selective agent against gram-negative bacteria in selective media for the detection of mollicutes. It is also used in preparation of thallium derivatives. It is involved in the separation ore constituents by flotation method.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 131°C |
Physical Form | Wet Crystalline Aggregates |
Quantity | 25 g |
UN Number | UN1707 |
Beilstein | 3693914 |
Merck Index | 14,9257 |
Solubility Information | Soluble in water,alcoholSoluble in cold water,ethanol,chloroform and ether; Sparingly soluble in hot water; Insoluble in acetone. |
Formula Weight | 263.42 |
Percent Purity | 99.995% |
Odor | Odorless |
Show More |
Safety and Handling
Hazard Category | H300+H330-H373 |
Hazard Statement | GHS H Statement H300-H330-H373 Fatal if swallowed. Fatal if inhaled. May cause damage to organs through prolonged or repeated exposure. |
Precautionary Statement | P260-P264b-P270-P271-P284-P301+P310-P304+P340-P310-P314-P330-P501c |
DOTInformation | Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: THALLIUM COMPOUNDS, N.O.S. |
EINECSNumber | 209-257-5 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.