Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Thallium(I) acetate, Puratronic™, 99.9985% (metals basis)

Manufacturer:  Alfa Aesar™ 1110222

Quantity 100g

 View more versions of this product

Catalog No. AA1110222


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

563-68-8
MFCD00013045
3693914
Thallous acetate
Puratronic™
263.42
UN1707
14,9257
Thallium(I) acetate

Specifications

563-68-8
263.42
UN1707
14,9257
Odorless
131°C
Thallium(I) acetate
Puratronic™
Powder
MFCD00013045
3693914
Thallous acetate
Hygroscopic
100g
99.9985%, (metals basis)

Safety and Handling

SDS & Safety Documents

GHS H Statement
H300-H330-H373
Fatal if swallowed.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.

GHS P Statement
P260-P301+P310-P304+P340-P320-P330-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: THALLIUM COMPOUNDS, N.O.S.

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit