Alfa Aesar™ Thallium(I) acetate, Puratronic™, 99.9985% (metals basis)

Manufacturer: Alfa Aesar™ 1110214

Quantity 25g

 View more versions of this product

Catalog No. AA1110214

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 563-68-8
    Formula Weight 263.42
    MDL Number MFCD00013045
    UN Number UN1707
    Beilstein 3693914
    Merck Index 14,9257
    Synonym Thallous acetate
    Chemical Name or Material Thallium(I) acetate
    Grade Puratronic™

    Specifications

    Formula Weight 263.42
    Health Hazard 3 GHS P Statement
    P260-P301+P310-P304+P340-P320-P330-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 2 GHS H Statement
    H300-H330-H373
    Fatal if swallowed.
    Fatal if inhaled.
    May cause damage to organs through prolonged or repeated exposure.
    TSCA TSCA
    Physical Form Powder
    UN Number UN1707
    Beilstein 3693914
    Synonym Thallous acetate
    Melting Point 131°C
    DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: THALLIUM COMPOUNDS, N.O.S.
    Sensitivity Hygroscopic
    MDL Number MFCD00013045
    Merck Index 14,9257
    Health Hazard 1 Danger
    Odor Odorless
    CAS 563-68-8
    Quantity 25g
    Chemical Name or Material Thallium(I) acetate
    Assay 99.9985%, (metals basis)
    Grade Puratronic™

    GHS H Statement
    H300-H330-H373
    Fatal if swallowed.
    Fatal if inhaled.
    May cause damage to organs through prolonged or repeated exposure.

    GHS P Statement
    P260-P301+P310-P304+P340-P320-P330-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit