Alfa Aesar™ Thallium(I) chloride, 99.999% (metals basis)

Manufacturer: Alfa Aesar™ 1183922

Quantity 100g

 View more versions of this product

Catalog No. AA1183922

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7791-12-0
    Formula Weight 239.82
    MDL Number MFCD00011274
    UN Number UN1707
    Merck Index 14,9260
    Synonym Thallous chloride
    Chemical Name or Material Thallium(I) chloride

    Specifications

    Formula Weight 239.82
    Health Hazard 3 GHS P Statement
    P260-P301+P310-P304+P340-P320-P330-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 2 GHS H Statement
    H300-H330-H373
    Fatal if swallowed.
    Fatal if inhaled.
    May cause damage to organs through prolonged or repeated exposure.
    DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: THALLIUM COMPOUNDS, N.O.S.
    Physical Form Powder
    Refractive Index 2.247
    Quantity 100g
    Density 7.0g/mL
    TSCA TSCA
    Sensitivity Air Sensitive
    MDL Number MFCD00011274
    UN Number UN1707
    Merck Index 14,9260
    Synonym Thallous chloride
    Health Hazard 1 Danger
    CAS 7791-12-0
    Melting Point 430°C
    RTECS Number XG4200000
    Chemical Name or Material Thallium(I) chloride
    Assay 99.999%, (metals basis)

    GHS H Statement
    H300-H330-H373
    Fatal if swallowed.
    Fatal if inhaled.
    May cause damage to organs through prolonged or repeated exposure.

    GHS P Statement
    P260-P301+P310-P304+P340-P320-P330-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit