Alfa Aesar™ Thallium(I) ethoxide, 98%

Manufacturer: Alfa Aesar™ 7813114

Quantity 25g

 View more versions of this product

Catalog No. AA7813114

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 20398-06-5
    Molecular Formula C2H5OTl
    Linear Formula TlOCH2CH3
    Formula Weight 249.43
    MDL Number MFCD00009072
    UN Number UN2810
    Chemical Name or Material Thallium (I) ethoxide

    Specifications

    Formula Weight 249.43
    Health Hazard 3 GHS P Statement
    P273-P309-P310-P302+P352-P420a-P501a
    Avoid release to the environment.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN: Wash with plenty of soap and water.
    Store away from other materials.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H300-H330-H373
    Fatal if swallowed.
    Fatal if inhaled.
    May cause damage to organs through prolonged or repeated exposure.
    Molecular Formula C2H5OTl
    CAS 20398-06-5
    Quantity 25g
    Flash Point >110°C (230°F)
    TSCA TSCA
    Refractive Index 1.6714
    Sensitivity Moisture Sensitive
    Linear Formula TlOCH2CH3
    MDL Number MFCD00009072
    UN Number UN2810
    Boiling Point 130°C (dec.)
    Melting Point −3°C
    Density 3.493g/mL
    RTECS Number KM2940000
    Physical Form Liquid
    DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
    Chemical Name or Material Thallium (I) ethoxide
    Assay 98%

    GHS H Statement
    H300-H330-H373
    Fatal if swallowed.
    Fatal if inhaled.
    May cause damage to organs through prolonged or repeated exposure.

    GHS P Statement
    P273-P309-P310-P302+P352-P420a-P501a
    Avoid release to the environment.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN: Wash with plenty of soap and water.
    Store away from other materials.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit