Alfa Aesar™ Thallium(I) iodide, 99.999% (metals basis)

Manufacturer: Alfa Aesar™ 1182514

Quantity 25g

 View more versions of this product

Catalog No. AA1182514

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 7790-30-9
    Formula Weight 331.27
    MDL Number MFCD00011279
    UN Number UN1707
    Merck Index 14,9264
    Chemical Name or Material Thallium(I) iodide

    Specifications

    Formula Weight 331.27
    Health Hazard 3 GHS P Statement
    P260-P301+P310-P304+P340-P320-P330-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Odor Odorless
    Quantity 25g
    Health Hazard 2 GHS H Statement
    H300-H330-H373
    Fatal if swallowed.
    Fatal if inhaled.
    May cause damage to organs through prolonged or repeated exposure.
    MDL Number MFCD00011279
    UN Number UN1707
    Density 7.1g/mL
    DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: THALLIUM COMPOUNDS, N.O.S.
    TSCA TSCA
    Physical Form Powder
    Merck Index 14,9264
    Boiling Point 824°C
    Solubility Information Practically insoluble in water
    Health Hazard 1 Danger
    CAS 7790-30-9
    Color Yellow
    Melting Point 440°C
    RTECS Number XG5425000
    Chemical Name or Material Thallium(I) iodide
    Assay 99.999%, (metals basis)

    GHS H Statement
    H300-H330-H373
    Fatal if swallowed.
    Fatal if inhaled.
    May cause damage to organs through prolonged or repeated exposure.

    GHS P Statement
    P260-P301+P310-P304+P340-P320-P330-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit