Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thallium(I) nitrate, 99.99%, (trace metal basis), Acros Organics™

Click to view available options
Quantity:
10g
Packaging:
Glass bottle
10102-45-1
NO3Tl
266.38
FYWSTUCDSVYLPV-UHFFFAOYSA-N
thallium i nitrate,thallium 1+ nitrate,thallous nitrate,thallium mononitrate,thallium nitrate van,thallium nitrate, tlno3,unii-ljq38dsr12,rcra waste number u217,thallium i nitrate 1:1
24937
thallium(1+);nitrate
[N+](=O)([O-])[O-].[Tl+]
10102-45-1
NO3Tl
266.38
FYWSTUCDSVYLPV-UHFFFAOYSA-N
thallium i nitrate,thallium 1+ nitrate,thallous nitrate,thallium mononitrate,thallium nitrate van,thallium nitrate, tlno3,unii-ljq38dsr12,rcra waste number u217,thallium i nitrate 1:1
24937
thallium(1+);nitrate
[N+](=O)([O-])[O-].[Tl+]
for ultra trace analysis
Assay Percent Range 75.9 to 77.5% (Tl)
Linear Formula TlNO3
Fieser 13,86
Merck Index 15,9414
Solubility Information Solubility in water: 95g/L (20°C). Other solubilities: insoluble in alcohol
Formula Weight 266.38
Physical Form Powder or Cyrstals
Percent Purity 99.99%
Grade Trace Metal Basis
Packaging Glass bottle
Solubility Solubility in water: 95g/L (20°C). Other solubilities: insoluble in alcohol
Total Trace Metal Impurities 100ppm max
Boiling Point 433.0°C
Color White
Melting Point 206.0°C
Quantity 10g
Chemical Name or Material Thallium(I) nitrate
Show More Show Less

GHS H Statement
May cause damage to organs through prolonged or repeated exposure.
Fatal if inhaled.
Toxic to aquatic life with long lasting effects.
May intensify fire; oxidizer.
Fatal if swallowed.

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Immediately call a POISON CENTER or doctor/physician.

GHS Signal Word: Danger

EINECSNumber : 233-273-1