Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Thallium(I) sulfate, Puratronic™, 99.9995% (metals basis)

Manufacturer:  Alfa Aesar™ 4021514

Quantity 25g

 View more versions of this product

Catalog No. AA4021514

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

7446-18-6
MFCD00011278
14,9269
Thallium(I) sulfate
504.8
UN1707
Thallous sulfate
Puratronic™

Specifications

7446-18-6
504.8
UN1707
Thallous sulfate
Odorless
25g
99.9995%, (metals basis)
Crystalline
MFCD00011278
14,9269
6.77g/mL
632°C
Thallium(I) sulfate
Puratronic™

Safety and Handling

SDS & Safety Documents

GHS H Statement
H300-H372-H315
Fatal if swallowed.
Causes damage to organs through prolonged or repeated exposure.
Causes skin irritation.

GHS P Statement
P260-P280-P301+P310-P362-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Take off contaminated clothing and wash before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: THALLIUM COMPOUNDS, N.O.S.

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit