Alfa Aesar™ Thallium(I) sulfate, Puratronic™, 99.9995% (metals basis)

Manufacturer: Alfa Aesar™ 4021506

Quantity 5g

 View more versions of this product

Catalog No. AA4021506

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 7446-18-6
    Formula Weight 504.8
    MDL Number MFCD00011278
    UN Number UN1707
    Merck Index 14,9269
    Synonym Thallous sulfate
    Chemical Name or Material Thallium(I) sulfate
    Grade Puratronic™

    Specifications

    Formula Weight 504.8
    Health Hazard 3 GHS P Statement
    P260-P280-P301+P310-P362-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Take off contaminated clothing and wash before reuse.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Physical Form Crystalline
    Melting Point 632°C
    Density 6.77g/mL
    TSCA TSCA
    Odor Odorless
    MDL Number MFCD00011278
    UN Number UN1707
    Merck Index 14,9269
    CAS 7446-18-6
    Synonym Thallous sulfate
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H300-H372-H315
    Fatal if swallowed.
    Causes damage to organs through prolonged or repeated exposure.
    Causes skin irritation.
    DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: THALLIUM COMPOUNDS, N.O.S.
    Quantity 5g
    Chemical Name or Material Thallium(I) sulfate
    Assay 99.9995%, (metals basis)
    Grade Puratronic™

    GHS H Statement
    H300-H372-H315
    Fatal if swallowed.
    Causes damage to organs through prolonged or repeated exposure.
    Causes skin irritation.

    GHS P Statement
    P260-P280-P301+P310-P362-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Take off contaminated clothing and wash before reuse.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit