Alfa Aesar™ Thallium(III) nitrate trihydrate, 99.5% (metals basis)

Manufacturer: Alfa Aesar™ 8799914

Quantity 25g

 View more versions of this product

Catalog No. AA8799914

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 13453-38-8
    Molecular Formula Tl(NO3)3·3H2O
    Formula Weight 444.43 (390.38 Anhydrous)
    MDL Number MFCD00149874
    UN Number UN2727
    Chemical Name or Material Thallium (III) nitrate trihydrate

    Specifications

    Formula Weight 444.43 (390.38 Anhydrous)
    Health Hazard 3 GHS P Statement
    P221-P301+P310-P304+P340-P320-P330-P405-P501a
    Take any precaution to avoid mixing with combustibles.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Odor Odorless
    Molecular Formula Tl(NO3)3·3H2O
    CAS 13453-38-8
    UN Number UN2727
    Health Hazard 2 GHS H Statement
    H300-H330-H272-H373
    Fatal if swallowed.
    Fatal if inhaled.
    May intensify fire
    oxidizer.
    May cause damage to organs through prolonged or repeated exposure.
    MDL Number MFCD00149874
    Quantity 25g
    DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: THALLIUM NITRATE
    RTECS Number QV2000000
    Physical Form Crystalline
    Sensitivity Hygroscopic
    Melting Point 102° to 105°C
    TSCA TSCA
    Health Hazard 1 Danger
    Chemical Name or Material Thallium (III) nitrate trihydrate
    Assay 99.5%, (metals basis)

    GHS H Statement
    H300-H330-H272-H373
    Fatal if swallowed.
    Fatal if inhaled.
    May intensify fire
    oxidizer.
    May cause damage to organs through prolonged or repeated exposure.

    GHS P Statement
    P221-P301+P310-P304+P340-P320-P330-P405-P501a
    Take any precaution to avoid mixing with combustibles.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit