Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Thallium(III) nitrate trihydrate, 99.5% (metals basis)

Manufacturer:  Alfa Aesar™ 8799922

Quantity 100g

 View more versions of this product

Catalog No. AA8799922


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

13453-38-8
444.43 (390.38 Anhydrous)
UN2727
Tl(NO3)3·3H2O
MFCD00149874
Thallium (III) nitrate trihydrate

Specifications

13453-38-8
Tl(NO3)3·3H2O
MFCD00149874
Odorless
102° to 105°C
Thallium (III) nitrate trihydrate
Crystalline
444.43 (390.38 Anhydrous)
UN2727
Hygroscopic
100g
99.5%, (metals basis)

Safety and Handling

SDS & Safety Documents

GHS H Statement
H300-H330-H272-H373
Fatal if swallowed.
Fatal if inhaled.
May intensify fire
oxidizer.
May cause damage to organs through prolonged or repeated exposure.

GHS P Statement
P221-P301+P310-P304+P340-P320-P330-P405-P501a
Take any precaution to avoid mixing with combustibles.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: THALLIUM NITRATE

RTECSNumber : QV2000000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit