Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Thallium(III) oxide, 99.9% (metals basis)

In organic synthesis and as an analytical reagent

Manufacturer:  Alfa Aesar™ 4379709

Quantity 10g

 View more versions of this product

Catalog No. AA4379709


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

1314-32-5
456.74
UN1707
Thallium sesquioxide; Thallic oxide
Tl2O3
MFCD00011276
14,9268
Thallium (III) oxide

Specifications

1314-32-5
Tl2O3
MFCD00011276
14,9268
Insoluble in water and alkalies; Decomposes in HCl and H2SO4
Odorless
717°C
Thallium (III) oxide
Powder
456.74
UN1707
Thallium sesquioxide; Thallic oxide
9.65g/mL
875°C
10g
99.9%, (metals basis)

Safety and Handling

SDS & Safety Documents

GHS H Statement
H300-H330-H373
Fatal if swallowed.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.

GHS P Statement
P260-P301+P310-P304+P340-P320-P330-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: THALLIUM COMPOUNDS, N.O.S.

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit