Alfa Aesar™ Thallium(III) oxide, 99.9% (metals basis)

In organic synthesis and as an analytical reagent

Manufacturer: Alfa Aesar™ 4379718

Quantity 50g

 View more versions of this product

Catalog No. AA4379718

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1314-32-5
    Molecular Formula Tl2O3
    Formula Weight 456.74
    MDL Number MFCD00011276
    UN Number UN1707
    Merck Index 14,9268
    Synonym Thallium sesquioxide; Thallic oxide
    Chemical Name or Material Thallium (III) oxide

    Specifications

    Formula Weight 456.74
    Health Hazard 3 GHS P Statement
    P260-P301+P310-P304+P340-P320-P330-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Molecular Formula Tl2O3
    DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: THALLIUM COMPOUNDS, N.O.S.
    TSCA TSCA
    UN Number UN1707
    Synonym Thallium sesquioxide; Thallic oxide
    Boiling Point 875°C
    Quantity 50g
    Solubility Information Insoluble in water and alkalies; Decomposes in HCl and H2SO4
    Health Hazard 1 Danger
    Odor Odorless
    CAS 1314-32-5
    Merck Index 14,9268
    Melting Point 717°C
    Health Hazard 2 GHS H Statement
    H300-H330-H373
    Fatal if swallowed.
    Fatal if inhaled.
    May cause damage to organs through prolonged or repeated exposure.
    Density 9.65g/mL
    Physical Form Powder
    MDL Number MFCD00011276
    Chemical Name or Material Thallium (III) oxide
    Assay 99.9%, (metals basis)

    GHS H Statement
    H300-H330-H373
    Fatal if swallowed.
    Fatal if inhaled.
    May cause damage to organs through prolonged or repeated exposure.

    GHS P Statement
    P260-P301+P310-P304+P340-P320-P330-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit