Alfa Aesar™ Thallium(III) trifluoroacetate, tech.

Manufacturer: Alfa Aesar™ 4364309

Quantity 10g

 View more versions of this product

Catalog No. AA4364309

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 23586-53-0
    Molecular Formula Tl(OOCCF3)3
    Formula Weight 543.42
    MDL Number MFCD00000414
    UN Number UN1707
    Synonym Thallic trifluoroacetate
    Chemical Name or Material Thallium (III) trifluoroacetate
    Grade Technical Grade

    Specifications

    Formula Weight 543.42
    Health Hazard 3 GHS P Statement
    P260-P301+P310-P304+P340-P320-P330-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Molecular Formula Tl(OOCCF3)3
    UN Number UN1707
    Synonym Thallic trifluoroacetate
    Melting Point 213°C (dec.)
    Physical Form Powder
    CAS 23586-53-0
    Health Hazard 2 GHS H Statement
    H300-H330-H373
    Fatal if swallowed.
    Fatal if inhaled.
    May cause damage to organs through prolonged or repeated exposure.
    TSCA TSCA
    Sensitivity Hygroscopic
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: THALLIUM COMPOUNDS, N.O.S.
    MDL Number MFCD00000414
    Quantity 10g
    Chemical Name or Material Thallium (III) trifluoroacetate
    Grade Technical Grade

    GHS H Statement
    H300-H330-H373
    Fatal if swallowed.
    Fatal if inhaled.
    May cause damage to organs through prolonged or repeated exposure.

    GHS P Statement
    P260-P301+P310-P304+P340-P320-P330-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit