Thallium Rod, 12.7mm (0.5in) dia., 99.9% (Metals basis), Alfa Aesar™

Form: Approx. 225g/150mm

Manufacturer: Alfa Aesar 123520Y

Quantity 75mm

 View more versions of this product

Catalog No. AA123520Y

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7440-28-0
    Physical Form Rod, 12.7mm (0.5 in.) dia.
    Assay Percent Range 99.9% (Metals basis)
    MDL Number MFCD00134063
    Chemical Name or Material Thallium

    Specifications

    Quantity 75mm
    EINECS Number 231-138-1
    Physical Form Rod, 12.7mm (0.5 in.) dia.
    CAS 7440-28-0
    Odor Odorless
    Appearance Rod
    Assay Percent Range 99.9% (Metals basis)
    MDL Number MFCD00134063
    Health Hazard 1 H300-H330-H373-H413: Fatal if swallowed. Fatal if inhaled. May cause damage to organs through prolonged or repeated exposure. May cause long lasting harmful effects to aquatic life.
    Chemical Name or Material Thallium

    H300-H330-H373-H413: Fatal if swallowed. Fatal if inhaled. May cause damage to organs through prolonged or repeated exposure. May cause long lasting harmful effects to aquatic life.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit