Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ THAP1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA567164
Description
Immunogen sequence: VNLSVFCDHNY TVEDTMHQRK RIHQLEQQVE KLRKKLKTAQ QRCRRQERQL EKLKEVVHFQ KEKDDVSERG YVILPNDYF Highest antigen sequence identity to the following orthologs - mouse 95%, rat 94%.
DNA-binding transcription regulator that regulates endothelial cell proliferation and G1/S cell-cycle progression. Specifically binds the 5'-[AT]NTNN[GT]GGCA[AGT]-3' core DNA sequence and acts by modulating expression of pRB-E2F cell-cycle target genes, including RRM1. Component of a THAP1/THAP3-HCFC1-OGT complex that is required for the regulation of the transcriptional activity of RRM1. May also have pro-apoptopic activity by potentiating both serum-withdrawal and TNF-induced apoptosis.
Specifications
THAP1 | |
Polyclonal | |
Unconjugated | |
Thap1 | |
4833431A01Rik; AW490810; DYT6; nuclear proapoptotic factor; THAP domain containing 1; THAP domain containing, apoptosis associated protein 1; THAP domain protein 1; THAP domain-containing apoptosis-associated protein 1; THAP domain-containing protein 1; THAP1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55145 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9NVV9 | |
Thap1 | |
Recombinant Human THAP1. Recombinant protein control fragment (Product #RP-107813). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction