Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
THAP2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA550803
Description
The antibody detects endogenous levels of total THAP2 protein.
THAP2 Polyclonal antibody specifically detects THAP2 in Human samples. It is validated for Immunohistochemistry (Paraffin).Specifications
THAP2 | |
Polyclonal | |
Unconjugated | |
THAP2 | |
2900040O07Rik; 9030625G08Rik; AI450385; AI649097; THAP domain containing 2; THAP domain containing, apoptosis associated protein 2; THAP domain-containing protein 2; Thap2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
83591 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.8 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q9H0W7 | |
THAP2 | |
Full length fusion protein of Human THAP2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction