Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ THAP3 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA550841

Catalog No. PA550841


Only null left
Add to Cart

Description

Description

The antibody detects endogenous levels of total THAP3 protein.

THAP3 (THAP domain-containing protein 3) is a 239 amino acid protein that contains one THAP-type zinc finger and exists as three alternatively spliced isoforms. The gene that encodes THAP3 contains roughly 10,721 bases and maps to human chromosome 1p36.31. Spanning about 260 million base pairs and comprising nearly 8% of the human genome, chromosome 1 is the largest human chromosome.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

THAP3
Polyclonal
Unconjugated
THAP3
2010013E08Rik; 2210418H06Rik; RGD1308109; snoRNA MBI-15; THAP domain containing 3; THAP domain containing, apoptosis associated protein 3; THAP domain-containing protein 3; THAP3
Rabbit
Antigen affinity chromatography
RUO
90326
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
1.8 mg/mL
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Q8WTV1
THAP3
Synthetic peptide corresponding to residues near the C terminal of human THAP domain containing, apoptosis associated protein 3.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.