Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

THAP8 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548531

Catalog No. PA548531

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

THAP8
Polyclonal
PBS with 0.09% sodium azide
Q8NA92
THAP domain-containing protein 8, THAP8
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
199745
Western Blot
Unconjugated
THAP8
Liquid
THAP8
KLH conjugated synthetic peptide between 72-100 amino acids from the Central region of human THAP8.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit