Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

THAP8 Mouse anti-Human, Clone: OTI5D11, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA527464

Catalog No. PIMA527464


Add to cart

Description

Description

THAP8 is a protein coding gene.
Specifications

Specifications

THAP8
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q8NA92
THAP domain-containing protein 8, THAP8
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
199745
Immunohistochemistry (Paraffin), Western Blot
OTI5D11
Unconjugated
THAP8
Liquid
THAP8
Human recombit protein fragment corresponding to amino acids 86-274 of THAP8 produced in E.coli
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit