Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

THBD Mouse anti-Human, Clone: THBD/1591, Abnova™

Mouse Monoclonal Antibody

Manufacturer:  Abnova Corporation MAB15007

Catalog No. 89-035-027


Add to cart

Description

Description

Mouse monoclonal antibody raised against partial recombinant human THBD.

  • High specificity
  • High reactivity
Specifications

Specifications

THBD
THBD/1591
Mouse monoclonal antibody raised against partial recombinant human THBD.
In 10 mM PBS.
Liquid
THBD
Recombinant protein corresponding to amino acids 69-194 of human THBD.
Protein A/G purification
RUO
Primary
7056
ELISA, Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
ELISA (2-4 ug/mL for coating) Flow Cytometry (0.5-1 ug/106 cells) Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1-2 ug/mL) Western Blotting (0.5-1 ug/mL) The optimal workin
P07204
CD141|THRM|TM
Mouse
IgG2b, kappa
100 ug
Store at -20 to -80°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only (RUO)