Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thrombospondin 3 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549120

Catalog No. PA549120

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Predicted to react with mouse based on sequence homology.

Adhesive glycoprotein that mediates cell-to-cell and cell-to-matrix interactions. Can bind to fibrinogen, fibronectin, laminin and type V collagen.
Specifications

Specifications

Thrombospondin 3
Polyclonal
Unconjugated
THBS3
Liquid
THBS3
KLH conjugated synthetic peptide between 227-255 amino acids from the N-terminal region of human THBS3.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Western Blot
0.4 mg/mL
PBS with 0.09% sodium azide
P49746
Thrombospondin-3, THBS3, TSP3
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
7059
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit