Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ PELTOR™ CH-3 Listen Only Hearing Protector

PELTOR™ CH-3 Listen Only Hearing Protector helps workers stay protected in noisy environments while enabling them to hear incoming audio from handheld devices.

$145.05 - $165.53

Specifications

ANSI S3.1974 Yes
Color Bright Yellow
For Use With (Application) Manufacturing
Product Type CH-3 Listen Only
View More Specs

Products 3
Catalog Number Mfr. No. Model Style Noise Reduction Rating (NRR) Price Quantity & Availability  
Catalog Number Mfr. No. Model Style Noise Reduction Rating (NRR) Price Quantity & Availability  
19-072-277
Accessories View Documents
3M™
7100203299
HT52A-112 Over-the-head 26 dB
Each for $147.30
Case of 10 EA for $1,473.00
Add to cart
 
19-072-249
Accessories View Documents
3M™
7100203298
HT52B-112 Neckband 25 dB
Each for $165.53
Case of 10 EA for $1,655.30
Add to cart
 
19-072-263
Accessories View Documents
3M™
7100203302
HT52P3E-112 Hard Hat 25 dB
Each for $145.05
Case of 10 EA for $1,450.50
Add to cart
 
Description

Description

  • Hearing protection solution with clear audio reception in noisy environments
  • FLX2 connection jack for easy and reliable connection to a wide assortment of handheld devices
  • Lasting comfort for long duration wear times
  • Designed to withstand harsh environmental elements
  • Audio speakers are optimized for speech frequencies and meet the efficiency demands of many of today's digital devices
  • Bright yellow color for enhanced visibility

Compliance

3M™ ear muffs must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.

Specifications

Specifications

Yes
Bright Yellow
Manufacturing
CH-3 Listen Only
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit