Learn More
THEG 293T Cell Overexpression Lysate (Denatured), Abnova
Manufacturer:  Abnova Corporation H00051298T01
Catalog No. 89-002-219
Add to cart
Description
Description
Western Blotting
Specifications
Specifications
Western Blot
293T
BC028574
6.8
Transient overexpression cell Lysate was tested with Anti-THEG antibody (H00051298-B01) by western blotting
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Human
THEG 293T Cell Transient Overexpression Lysate(Denatured)
41.8
pCMV-THEG full-length
100 uL
Store at -80°C. Aliquot to avoid repeated freezing and thawing.