Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

THEG Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP17969320UL

 View more versions of this product

Catalog No. NBP17969320


Add to cart

Description

Description

THEG Polyclonal antibody specifically detects Antigen in Human samples. It is validated for Western Blotting.
Specifications

Specifications

THEG
Unconjugated
PBS and 2% Sucrose with 0.09% Sodium Azide
Affinity Purified
THEG
Synthetic peptide directed towards the middle region of human THEG. Peptide sequence: DRPSVYWTERFLEDTTLTITVPAVSRRVEELSRPKRFYLEYYNNNRTTPV
41 kDa
20ul
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Human
Western Blot
Western Blot 1:1000
NP_954672
CT56Cancer/testis antigen 56, MGC26138, testis-specific, Theg homolog, Theg homolog (mouse)
Rabbit
IgG
Immunogen affinity purified
RUO
Primary
51298
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit