Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Themis Rabbit anti-Human, Mouse, FITC, Polyclonal, FabGennix
Rabbit Polyclonal Antibody
Supplier: Fabgennix Inc THMSFITC
Description
Themis Polyclonal antibody specifically detects Themis in Human, Mouse samples. It is validated for ELISA, Immunohistochemistry, Immunoprecipitation, Western BlotSpecifications
Themis | |
Polyclonal | |
FITC | |
Themis | |
bA325O24.3; bA325O24.4; C6orf190; C6orf207; E430004N04Rik; FLJ40584; Gasp; GRB2-associated protein; Grb2-associating protein; MGC163388; Protein THEMIS; Protein thylex; RGD1560849; signaling phosphoprotein specific for T cells; SPOT; THEMIS; thylex; thymocyte expressed molecule involved in selection; thymocyte selection associated; thymocyte selection pathway associated; thymocyte-expressed molecule involved in selection; Tsepa | |
Synthetic peptide taken within amino acid region 1-50 on Thymocyte-expressed molecule involved in selection protein. | |
200 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot | |
0.5-1.5 mg/mL | |
proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8 | |
Q8BGW0, Q8N1K5 | |
Rabbit | |
Affinity chromatography | |
RUO | |
210757, 387357 | |
-20° C, store in dark | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction