Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Theobromine, EP, 97-101%, Spectrum™

Click to view available options
Quantity:
10 kg
25 kg
Packaging:
Fiber Drum
Poly Pail
83-67-0
C7H8N4O2
180.17
YAPQBXQYLJRXSA-UHFFFAOYSA-N
3,7-dimethyl-2,3,6,7-tetrahydro-1H-purine-2,6-dione
CN1C=NC2=C1C(=O)NC(=O)N2C
83-67-0
C7H8N4O2
180.17
YAPQBXQYLJRXSA-UHFFFAOYSA-N
3,7-dimethyl-2,3,6,7-tetrahydro-1H-purine-2,6-dione
CN1C=NC2=C1C(=O)NC(=O)N2C

T3960, 83-67-0, C7H8N4O2

Spectrum™ Theobromine, EP
Packaging Poly Pail
Sulfated Ash 0.001
Loss on Drying 0.005
Quantity 10 kg
CAS Max % 1
Percent Purity 97 to 101%
Grade EP