Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Theophylline, Anhydrous, USP, 97-102%, Spectrum™

Click to view available options
Quantity:
100 g
500 g
2.5 kg
12 kg
25 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
58-55-9
C7H8N4O2
180.17
ZFXYFBGIUFBOJW-UHFFFAOYSA-N
1,3-dimethyl-2,3,6,7-tetrahydro-1H-purine-2,6-dione
CN1C2=C(NC=N2)C(=O)N(C)C1=O
58-55-9
C7H8N4O2
180.17
ZFXYFBGIUFBOJW-UHFFFAOYSA-N
1,3-dimethyl-2,3,6,7-tetrahydro-1H-purine-2,6-dione
CN1C2=C(NC=N2)C(=O)N(C)C1=O

TH110, 58-55-9, C7H8N4O2

Spectrum™ Theophylline, Anhydrous, USP is an oral medication used to treat asthma and other lung diseases. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Packaging Fiber Drum
Loss on Drying 0.005
Ignition Residue 0.0015
Quantity 25 kg
CAS Max % 1
Percent Purity 97 to 102%
Grade USP