Therapak™ 24-Hour Urine Containers Prefilled with Preservative Solution

Used to preserve urine samples during collection and transport for subsequent testing and diagnosis of disease

Manufacturer: Therapak CorporationSupplier Diversity Partner 38533G

 View more versions of this product

Catalog No. 22-130-554

  Check Availability
Add to Cart

    Specifications

    Product Type 24 Hour Urine Containers
    Capacity (Metric) 3L
    Cap Type Extra thick liner cap
    Color Amber
    Material Polyethylene
    Quantity 24 Case
    For Use With (Application) Used to preserve Urine samples during collection and transport for subsequent testing and diagnosis of disease
    Includes 30mL Acetic Acid, Plastic cap with extra thick liner

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit