Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Therapak™ 95kPa Compliant Transport Bags

For use as a secondary water-tight receptacle

Manufacturer:  Therapak CorporationSupplier Diversity Partner 15655

 View more versions of this product

Catalog No. 22-130-023

  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

Two-pocket bag with Aqui-Pak pouch
11 in.
Clear with white tape liner
Plastic
11 x 7 in. (27.9 x 17.8cm)
7 in.
27.9cm
17.8cm
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit