Learn More
Therapak™ Coleman™ Excursion™ Handle Coolers
For safe transport of contents
Manufacturer:  Therapak™ 56231G
 View more versions of this product
Catalog No. 22-131-510
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • Generous foam insulation, along with hinged lids and lid-locking handles
  • Available in red and blue color
  • Capacity: 9 to 16 qt. (8.51 to 15.14L)
Specifications
Specifications
8.51 L
Blue
Excursion Cooler
Hinged