Learn More
Therapak™ Duramark™ Thermoelectric Cooler/Warmer
Active solution for the transport of laboratory medical specimens
Supplier: Therapak™ 562059G
Description
- Use of 12V power plug in vehicle, each unit will cool to approximately 40°F lower than ambient temperature or heat to around 135°F
- Optional equipment includes battery saver switch which protects the car's battery from premature draining
- AC Power adapter allows the unit to be used in any 110/120 volt AC wall outlet
Order Info
Battery, adapter and power cords available separately (Cat. Nos. 22-131-495, 22-131-491 and 22-131-494).
Specifications
43.8 cm | |
39.3 cm | |
41.2 cm | |
15.5 x 16.25 x 17.25 in. (39.3 x 41.2 x 43.8 cm) | |
13 lb. | |
Compact Cooler, 12V |
17.25 in. | |
15.5 in. | |
16.25 in. | |
For transport of laboratory medical specimens | |
5.89 kg | |
27.4 L |