Therapak™ Exempt Human Specimen Label

For use with regulated specimens that do not fit into Infectious Category A or Biological Substances Category B classification

Manufacturer: Therapak CorporationSupplier Diversity Partner 54814

 View more versions of this product

Catalog No. 22-130-070

  Check Availability
Add to Cart

    Specifications

    Format Label
    For Use With (Application) Used to transport biological specimens
    Product Type Exempt Human Specimen Label
    Quantity 500 Pack
    Size (English) 2.5 x 2 in.
    Size (Metric) 6.35 x 5cm
    Certifications/Compliance IATA

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit